Learn More
Drummond™ Fixed-Volume Microdispensers
Catalog No. p-2363841
Click to view available options
Volume (Metric):
10 μL
100 μL
100 to 200 μL
20 μL
25 μL
250 μL
5 μL
50 μL
Overall volumetric accuracy of ±1% at full volume
  • Contain borosilicate glass bores with stainless steel plungers, except the 1000μL unit which has precision-molded plastic bores
  • All plungers with capacities 25μL and larger have PTFE tips
  • The 5μL unit (Cat. No. 21-170-10) will deliver 1 to 5μL
Order Info
Repeat Kit is available for 1000μL unit. Custom spacers are available permitting fractional deliveries on all units; contact your Fisher Scientific™ Customer Service Representative.
Specifications
Accuracy ±1%
Number of Channels 1
Type Fixed Volume Microdispenser
Volume (Metric) 5 μL
For Use With (Application) Precision molded Plastic bores
Includes 2 Plungers, 2 O-rings, 1 collet, 1 coil spring, 1 spring guide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.