Learn More
Drummond™ Dispensing Manifold
Catalog No. 2116910E
Click to view available options
Angle:
No
Yes
Number of Channels:
8
12
Eight-tip manifolds for use with the 21-169-10A Digital Microdispenser or any pipetter that accepts standard Luer-Lok™ connector
  • Stainless steel and PTFE construction
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection
Specifications
Angle Yes
Description 9mm (center to center); 11mm pin length
Material Stainless Steel, PTFE
Number of Channels 8
For Use With (Equipment) Drummond™ micropipetter
Includes Cleaning wire, Luer connector and end caps
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.