Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Drummond™ Wiretrol™ I Calibrated Micropipets
                    Click to view available options
                
                
                    
                            
                                Volume (Metric):
                            
                        
                        
                            
                                20 μL
                            
                        
                    Description
Includes
Handy dispenser cap, handle, stainless-steel plunger
- Deliver within 1%
 - One-handed operation
 - Accepts both high- and low-viscosity liquids including corrosives
 - Color coded
 - Micropipet 21-175D, -175E and -175F have PTFE-fluorocarbon–resin tips on plunger
 
Compliance
Calibrated To Contain
Specifications
Specifications
| Calibration | To Contain | 
| Material | Glass | 
| For Use With (Application) | For a wide variety of applications, Wiretrol™ will accept both high and low viscosity liquids including corrosives | 
| Includes | Handy dispenser cap, handle, Stainless Steel plunger | 
| Color | Black | 
| Volume (Metric) | 20 μL | 
| Type | Calibrated Micropipet | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction