Learn More
Drummond™ Wiretrol™ II Calibrated Micropipets

Description
Includes
Each case comes with handy dispenser cap, handle, and two stainless-steel plungers: one full length, plus one short length for noncontact delivery
Accept both high- and low-viscosity liquids, including corrosives
- Deliver within 1%
- One-handed operation
- Similar to Wiretrol I micropipets, except each pipet has two capacities, thus two easy-to-read volume lines (21-176-2A has six capacities, six lines) that won't wash off
- Micropipets 21-176-2D, 21-176-3A and 21-176-4A have PTFE-resin tips on plunger
Compliance
Calibrated To Contain.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Accuracy | ±1% |
| Calibration | To Contain |
| Material | Glass |
| For Use With (Application) | For a wide variety of applications, Wiretrol™ will accept both high and low viscosity liquids including corrosives |
| Includes | Handy dispenser cap, handle, two Stainless Steel plunger one full length, plus one short length for noncontact delivery, standard length and short length plungers |
| Color | Orange |
| Volume (Metric) | 10 and 20 μL |
| Type | Wiretrol™ II Calibrated Micropipet |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.