Learn More
Beckman Coulter Dry Powder System (DPS) HiCapacity HEPA Vacuum Unit and Accessories
The HiCap HEPA Vacuum Unit is designed for use with the Dry Powder System (DPS) of the LS 13 320 XR Particle Size Analyzer. The Vacuum Unit delivers dry powder samples to the sensing zone in the optical bench of the LS 13 320 XR, all required connectors are included.
Supplier:  Beckman Coulter C40017/DEL
 View more versions of this product
Catalog No. 15-655-210
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Description
Description
  • LS 13 320 XR Particle Size Analyzers equipped with the Dry Powder System (DPS) Module require a vacuum unit for use
  • HiCap HEPA Vacuum Units deliver dry powder samples to the sensing zone in the optical bench
  • Vacuums feature a fully adjustable pressure range
  • Replacement HiCap Vacuum Filters are available
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HiCap Vacuum Replacement Filter
LS 13 320 XR and Dry Powder System (DPS)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.