Learn More
Beckman Coulter Dry Powder System (DPS) HiCapacity HEPA Vacuum Unit and Accessories
The HiCap HEPA Vacuum Unit is designed for use with the Dry Powder System (DPS) of the LS 13 320 XR Particle Size Analyzer. The Vacuum Unit delivers dry powder samples to the sensing zone in the optical bench of the LS 13 320 XR, all required connectors are included.
Specifications
For Use With (Equipment) LS 13 320 XR and Dry Powder System (DPS)
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
15-655-210
View Documents
Beckman Coulter
C40017/DEL
HiCap Vacuum Replacement Filter N/A N/A N/A
Description
Description
  • LS 13 320 XR Particle Size Analyzers equipped with the Dry Powder System (DPS) Module require a vacuum unit for use
  • HiCap HEPA Vacuum Units deliver dry powder samples to the sensing zone in the optical bench
  • Vacuums feature a fully adjustable pressure range
  • Replacement HiCap Vacuum Filters are available
Specifications
Specifications
LS 13 320 XR and Dry Powder System (DPS)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.