Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DS2
GREENER_CHOICE

Catalog No. 367950R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-795-0R 50 mg
36-791-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-795-0R Supplier Tocris Bioscience™ Supplier No. 3679/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Positive allosteric modulator of δ-containing GABAA receptors

DS2 is a positive allosteric modulator of GABAA receptors. Potentiates GABA-evoked currents mediated by α4β3δ receptors (EC50 = 142 nM, in vitro). Displays complex selectivity profile, with selectivity for δ-containing receptors (α4/6βxδ > α1βxδ >> γ2 > α4β3 GABAA receptors). Enhances tonic currents of thalamocortical neurons in wild-type mice.

Specifications

Quantity 50 mg
Product Type DS2
Purity >97%
Molecular Weight (g/mol) 353.83
Inhibitors DS2

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.