Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ dsbC (Escherichia coli) Recombinant Protein

Catalog No. 89942442
Click to view available options
Quantity:
100 μg

Used for SDS-PAGE

Sequence: DDAAIQQTLAKMGIKSSDIQPAPVAGMKTVLTNSGVLYITDDGKHIIQGPMYDVSGTAPVNVTNKMLLKQLNALEKEMIVYKAPQEKHVITVFTDITCGYCHKLHEQMADYNALGITVRYLAFPRQGLDSDAEKEMKAIWCAKDKNKAFDDVMAGKSVAPASCDVDIADHYVLGVQLGVSGTPAVVLSNGTLVPGYQPPKEMKEFLDEHQKMTSGK

Specifications

Accession Number YP_003045913
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 8177108
Molecular Weight (g/mol) 21.1kDa
Name dsbC (Escherichia coli) Recombinant protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing 15% SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Immunogen DDAAIQQTLAKMGIKSSDIQPAPVAGMKTVLTNSGVLYITDDGKHIIQGPMYDVSGTAPVNVTNKMLLKQLNALEKEMIVYKAPQEKHVITVFTDITCGYCHKLHEQMADYNALGITVRYLAFPRQGLDSDAEKEMKAIWCAKDKNKAFDDVMAGKSVAPASCDVDIADHYVLGVQLGVSGTPAVVLSNGTLVPGYQPPKEMKEFLDEHQKMTSGK
Storage Requirements Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name DSBC
Gene Symbol NULL
Species E. coli
Protein Tag None
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.