Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584984 Supplier Invitrogen™ Supplier No. PA584984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQVILQVGVI NEAQFSKAAS SQTPTMCTTT VTVKIIDSDE GPECHPPVKV IQSQDGFPAG QELLGYKALD PEISSGEGLR YQKLGDEDNW FEINQHTG Highest antigen sequence indentity to the following orthologs: Rat - 77%, Mouse - 80%.

The desmosomal cadherin desmocollin DSC1 is expressed in upper epidermis where strong adhesion is required. DSC1 is a type I membrane protein (4) required for strong adhesion and barrier maintenance in epidermis and contributes to epidermal differentiation.The DSC1 gene comprises 17 exons spanning approximately 33 kb on 18q12.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DSC1
Gene Accession No. Q08554
Gene Alias 1110020A10Rik; AI507491; Cadherin family member 1; CDHF1; desmocollin 1; Desmocollin1; desmocollin-1; Desmosomal glycoprotein 2/3; DG2/DG3; DSC1; Dsc1a; Dsc1b
Gene Symbols DSC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DSC1. Recombinant protein control fragment (Product #RP-108273).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.