Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587603 Supplier Invitrogen™ Supplier No. PA587603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQDGFPAGQE LLGYKALDPE ISSGEGLRYQ KLGDEDNWFE INQHTGDLRT LKVLDRESKF VKNNQYNISV VAVDAVGRSC TGTLVVHLDD YNDHAPQIDK EVTICQNNED FAVLKPVDPD GPENGPPFQF FLDNSASKNW NIEEKDGKTA ILRQRQNLDY NYYSVPIQIK DRHGLVATHM LTVRVCDCST PSECRMKDKS TRDVRPNVIL G; Positive Samples: Raji, Mouse liver, Rat liver; Cellular Location: Cell junction, Cell membrane, Single-pass type I membrane protein, desmosome.

The desmosomal cadherin desmocollin DSC1 is expressed in upper epidermis where strong adhesion is required. DSC1 is a type I membrane protein (4) required for strong adhesion and barrier maintenance in epidermis and contributes to epidermal differentiation.The DSC1 gene comprises 17 exons spanning approximately 33 kb on 18q12.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DSC1
Gene Accession No. P55849, Q08554
Gene Alias 1110020A10Rik; AI507491; Cadherin family member 1; CDHF1; desmocollin 1; Desmocollin1; desmocollin-1; Desmosomal glycoprotein 2/3; DG2/DG3; DSC1; Dsc1a; Dsc1b
Gene Symbols DSC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 480-690 of human DSC1 (NP_0777391).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13505, 1823, 291759
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.