Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DSCAM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542698
Description
Peptide sequence: MRVCNSAGCA EKQANFATLN YDGSTIPPLI KSVVQNEEGL TTNEGLKMLV Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
This gene is a member of the immunoglobulin superfamily of cell adhesion molecules (Ig-CAMs), and is involved in human central and peripheral nervous system development. This gene is a candidate for Down syndrome and congenital heart disease (DSCHD). A gene encoding a similar Ig-CAM protein is located on chromosome 11. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Oct 2012].
Specifications
DSCAM | |
Polyclonal | |
Unconjugated | |
DSCAM | |
4932410A21Rik; CHD2; CHD2-42; CHD2-52; Down syndrome cell adhesion molecule; Down syndrome cell adhesion molecule homolog; DS cell adhesion molecule; Dscam; LOW QUALITY PROTEIN: Down syndrome cell adhesion molecule | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13508, 1826 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O60469, Q9ERC8 | |
DSCAM | |
Synthetic peptide directed towards the middle region of human DSCAM. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction