Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DSCAM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582674
Description
Immunogen sequence: PRAQLLIEER DTMETIDDRS TVLLTDADFG EAAKQKSLTV THTVHYQSVS QATGPLVDVS DARPGTNPTT RRNAKAGPTA RNRYASQWTL NRPHPTISAH TLTTDWRLPT PRAAGSVDKE SDSYSVSPSQ DTDRARSS.
This gene is a member of the immunoglobulin superfamily of cell adhesion molecules (Ig-CAMs), and is involved in human central and peripheral nervous system development. This gene is a candidate for Down syndrome and congenital heart disease (DSCHD). A gene encoding a similar Ig-CAM protein is located on chromosome 11. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Oct 2012].
Specifications
DSCAM | |
Polyclonal | |
Unconjugated | |
DSCAM | |
4932410A21Rik; CHD2; CHD2-42; CHD2-52; Down syndrome cell adhesion molecule; Down syndrome cell adhesion molecule homolog; DS cell adhesion molecule; Dscam; LOW QUALITY PROTEIN: Down syndrome cell adhesion molecule | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1826 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.25 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60469 | |
DSCAM | |
Recombinant protein corresponding to Human DSCAM. Recombinant protein control fragment (Product #RP-90470). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction