Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSCAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518559
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518559 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518559 Supplier Invitrogen™ Supplier No. PA518559
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene is a member of the immunoglobulin superfamily of cell adhesion molecules (Ig-CAMs), and is involved in human central and peripheral nervous system development. This gene is a candidate for Down syndrome and congenital heart disease (DSCHD). A gene encoding a similar Ig-CAM protein is located on chromosome 11. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Oct 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSCAM
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DSCAM
Gene Accession No. O60469
Gene Alias 4932410A21Rik; CHD2; CHD2-42; CHD2-52; Down syndrome cell adhesion molecule; Down syndrome cell adhesion molecule homolog; DS cell adhesion molecule; Dscam; LOW QUALITY PROTEIN: Down syndrome cell adhesion molecule
Gene Symbols DSCAM
Host Species Goat
Immunogen Synthetic peptide sequence (DSWDSAQRTKDVSPQ) corresponding to the internal amino acids of DSCAM (aa 927-942).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1826
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.