Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DSCAML1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA557201
Description
Immunogen sequence: LARTYHTQAR HLTLDPASKS LGLPHPGAPA AASTATLPQR TLAMPAPPAG TAPPAPGPTP AEPPTAPSAA PPAPSTEPPR AGGPHTKMGG SRDSLLEMST Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
DSCAML1 Polyclonal antibody specifically detects DSCAML1 in Human samples. It is validated for Immunohistochemistry.
Specifications
DSCAML1 | |
Polyclonal | |
Unconjugated | |
DSCAML1 | |
4921507G06Rik; 4930435C18Rik; BB239540; down syndrome cell adhesion molecule 2; Down syndrome cell adhesion molecule like 1; Down syndrome cell adhesion molecule-like 1; Down syndrome cell adhesion molecule-like protein 1; Down syndrome cell adhesion molecule-like protein 1 homolog; Downs syndrome cell adhesion molecule like 1; DS cell adhesion molecule like 1; DSCAM2; DSCAML1; DSCAM-like 1; KIAA1132; mKIAA1132 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57453 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TD84 | |
DSCAML1 | |
Recombinant protein corresponding to Human DSCAML1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction