Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DSCAML1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547476
Description
In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant human DSCAM is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
DSCAML1 encodes a protein that is a member of the Ig superfamily of cell adhesion molecules and is involved in neuronal differentiation. The encoded membrane-bound protein localizes to the cell surface, where it forms aggregates that repel neuronal processes of the same cell type.
Specifications
DSCAML1 | |
Polyclonal | |
Unconjugated | |
DSCAML1 | |
4921507G06Rik; 4930435C18Rik; BB239540; down syndrome cell adhesion molecule 2; Down syndrome cell adhesion molecule like 1; Down syndrome cell adhesion molecule-like 1; Down syndrome cell adhesion molecule-like protein 1; Down syndrome cell adhesion molecule-like protein 1 homolog; Downs syndrome cell adhesion molecule like 1; DS cell adhesion molecule like 1; DSCAM2; DSCAML1; DSCAM-like 1; KIAA1132; mKIAA1132 | |
Goat | |
Antigen Affinity Chromatography | |
RUO | |
57453 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8TD84 | |
DSCAML1 | |
Mouse myeloma cell line NS0-derived recombinant human DSCAM-L1 Glu19-Lys1591. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction