Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DSCC1 Antibody (V76P1B2*B12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$170.00
Specifications
Antigen | DSCC1 |
---|---|
Clone | V76P1B2*B12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Description
DSCC1 Monoclonal antibody specifically detects DSCC1 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
DSCC1 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Epigenetics | |
PBS | |
79075 | |
IgG | |
Protein A purified |
V76P1B2*B12 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DCC1MGC5528, defective in sister chromatid cohesion 1 homolog (S. cerevisiae), Defective in sister chromatid cohesion protein 1 homolog, hDCC1, sister chromatid cohesion protein DCC1 | |
Peptide Sequence - KVYNSRRPIS (immunogen peptide is identical in sheep, bovine, canine, equine, chicken and zebrafish) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title