Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DSCC1 Antibody (V76P1B2*B12), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250380V
Description
DSCC1 Monoclonal specifically detects DSCC1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
DSCC1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DSCC1 | |
Peptide Sequence - KVYNSRRPIS (immunogen peptide is identical in sheep, bovine, canine, equine, chicken and zebrafish) | |
0.1 mL | |
Epigenetics | |
79075 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
V76P1B2*B12 | |
Western Blot, ELISA, Immunohistochemistry | |
DCC1MGC5528, defective in sister chromatid cohesion 1 homolog (S. cerevisiae), Defective in sister chromatid cohesion protein 1 homolog, hDCC1, sister chromatid cohesion protein DCC1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction