Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554885 Supplier Invitrogen™ Supplier No. PA554885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGMVTSLDQL KGLALVDRHS RPEIIFLLKV DDLPEDNQER FNSLFSLREK WTEEDIAPYI QDLCGEKQTI GALLTKYSHS SMQNGVKVYN SRRP Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

CHTF18 (MIM 613201), CHTF8 (MIM 613202), and DSCC1 arecomponents of an alternative replication factor C (RFC) (see MIM600404) complex that loads PCNA (MIM 176740) onto DNA during Sphase of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSCC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DSCC1
Gene Accession No. Q9BVC3
Gene Alias 2010006I05Rik; 2600005O03Rik; DCC1; defective in sister chromatid cohesion 1 homolog; defective in sister chromatid cohesion 1 homolog (S. cerevisiae); defective in sister chromatid cohesion protein 1 homolog; DNA replication and sister chromatid cohesion 1; DSCC1; RGD1561749; Sister chromatid cohesion protein DCC1; UNQ9337/PRO34008
Gene Symbols DSCC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DSCC1. Recombinant protein control fragment (Product #RP-93796).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79075
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.