Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599006
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599006 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599006 Supplier Invitrogen™ Supplier No. PA599006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DSCC1 Polyclonal Antibody for Western Blot, ELISA

CHTF18 (MIM 613201), CHTF8 (MIM 613202), and DSCC1 arecomponents of an alternative replication factor C (RFC) (see MIM600404) complex that loads PCNA (MIM 176740) onto DNA during Sphase of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSCC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DSCC1
Gene Accession No. Q9BVC3
Gene Alias 2010006I05Rik; 2600005O03Rik; DCC1; defective in sister chromatid cohesion 1 homolog; defective in sister chromatid cohesion 1 homolog (S. cerevisiae); defective in sister chromatid cohesion protein 1 homolog; DNA replication and sister chromatid cohesion 1; DSCC1; RGD1561749; Sister chromatid cohesion protein DCC1; UNQ9337/PRO34008
Gene Symbols DSCC1
Host Species Rabbit
Immunogen Recombinant Human Sister chromatid cohesion protein DCC1 protein (1-393AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79075
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.