Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DSCR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554747
Description
Immunogen sequence: GKFPSGKTEI PFEFPLHLKG NKVLYETYHG VFVNIQYTLR CDMKRSLLAK DLTKTCEFIV HSAPQKGKFT PSPV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
The region of chromosome 21 between genes CBR and ERG (CBR-ERG region), which spans 2.5 Mb on 21q22.2, has been defined by analysis of patients with partial trisomy 21. It contributes significantly to the pathogenesis of many characteristics of Down syndrome, including morphological features, hypotonia, and mental retardation. The DSCR3 (Down syndrome critical region gene 3) gene is found in this region and is predictated to contain eight exons. DSCR3 is expressed in most tissues examined.
Specifications
DSCR3 | |
Polyclonal | |
Unconjugated | |
VPS26C | |
DCRA; Down syndrome critical region 3; Down syndrome critical region gene 3; Down syndrome critical region protein 3; Down syndrome critical region protein A; DSCR3; DSCR3 arrestin fold containing; DSCRA; Vacuolar protein sorting-associated protein 26C; VPS26C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10311 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14972 | |
VPS26C | |
Recombinant protein corresponding to Human DSCR3. Recombinant protein control fragment (Product #RP-91823). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction