Learn More
Invitrogen™ DSCR3 Polyclonal Antibody

Description
DSCR3 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | DSCR3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | VPS26C |
| Gene Accession No. | O14972 |
| Gene Alias | DCRA; Down syndrome critical region 3; Down syndrome critical region gene 3; Down syndrome critical region protein 3; Down syndrome critical region protein A; DSCR3; DSCR3 arrestin fold containing; DSCRA; Vacuolar protein sorting-associated protein 26C; VPS26C |
| Gene Symbols | VPS26C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.