Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DSCR9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144093
Description
DSCR9 Polyclonal Antibody for IHC (P), ELISA
DSCR9 is a gene that is found in the human genome. It is an example of a gene identifier or accession number, which is used to uniquely identify specific genes in genetic databases. However, without further context or information, it is difficult to provide specific details about the function or characteristics of DSCR9.
Specifications
DSCR9 | |
Polyclonal | |
Unconjugated | |
DSCR9 | |
DSCR9Down syndrome critical region protein 9 | |
Rabbit | |
Protein G | |
RUO | |
257203 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P59020 | |
DSCR9 | |
Recombinant Human Down syndrome critical region protein 9 protein (1-149AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction