Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dsDNA Antibody (DSD/4054R), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308490PCP
Description
dsDNA Monoclonal antibody specifically detects dsDNA in Human samples. It is validated for Flow CytometrySpecifications
dsDNA | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
DSD/4054R | |
Flow Cytometry | |
Double stranded DNA | |
Nuclei of Burkitt's cells | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction