Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dsDNA (Double strand-DNA) Monoclonal Antibody (121-3), Abbomax

Catalog No. 502204446
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4446 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4446 Supplier Abbomax Inc Supplier No. 606430
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Breast carcinoma Cellular location: Nucleus.

dsDNA (Double strand-DNA) Monoclonal antibody specifically detects dsDNA (Double strand-DNA) in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen dsDNA (Double strand-DNA)
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 121-3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Alias dsdna
Host Species Mouse
Immunogen Double-stranded DNA
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.