Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dsDNA Antibody (rDSD/4565) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119192 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119192 Supplier Novus Biologicals Supplier No. NBP308466
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

dsDNA Monoclonal specifically detects dsDNA in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen dsDNA
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rDSD/4565
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin : 1-2 ug/mL
Formulation 10mM PBS
Host Species Mouse
Immunogen Nuclei of Burkitt's cells
Purification Method Bioreactor concentrate
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.