Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSEL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566352
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566352 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566352 Supplier Invitrogen™ Supplier No. PA566352
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WTGEEVGDAAG EIITASQHGE MVFVSGEAVS AYSSAMRLKS VYRALLLLNS QTLLVVDHIE RQEDSPINSV SAFFHNLDID FKYIPYKFMN RYNGAMMDVW DA Highest antigen sequence identity to the following orthologs - mouse 94%, rat 92%.

DSEL gene ontology annotations related to this gene include chondroitin sulfate metabolic process; dermatan sulfate biosynthetic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSEL
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DSEL
Gene Accession No. Q8IZU8
Gene Alias C18orf4; dermatan sulfate epimerase-like; dermatan-sulfate epimerase-like protein; DSEL; NCAG1
Gene Symbols DSEL
Host Species Rabbit
Immunogen Recombinant Human DSEL. Recombinant protein control fragment (Product #RP-105160).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92126
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.