Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSG1 Mouse anti-Human, Clone: 18D4, Abnova™

Catalog No. 89034582 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-582 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-582 Supplier Abnova Corporation Supplier No. MAB14499
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human DSG1.

  • High specificity
  • High reactivity

Specifications

Antigen DSG1
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 18D4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human DSG1.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. Q02413
Gene Alias CDHF4/DG1/DSG
Gene Symbols DSG1
Host Species Mouse
Immunogen Recombinant protein corresponding to intracellular domain of human DSG1.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1828
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.