Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSG3 Mouse anti-Human, Clone: 5H10, Abnova™

Catalog No. 89034588 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-588 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-588 Supplier Abnova Corporation Supplier No. MAB14505
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human DSG3.

  • High specificity
  • High reactivity

Specifications

Antigen DSG3
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5H10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human DSG3.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P32926
Gene Alias CDHF6/DKFZp686P23184/PVA
Gene Symbols DSG3
Host Species Mouse
Immunogen Recombinant protein corresponding to extracellular domain of human DSG3.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1830
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.