Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DSN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | DSN1 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
DSN1 Polyclonal specifically detects DSN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DSN1 | |
Polyclonal | |
Rabbit | |
Human | |
C20orf172, chromosome 20 open reading frame 172, dJ469A13.2, DSN1, MIND kinetochore complex component, homolog (S. cerevisiae), FLJ13346, hKNL-3, kinetochore null 3 homolog, kinetochore-associated protein DSN1 homolog, KNL3, MIS13MGC32987 | |
DSN1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
79980 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KEYITKFSLERQTWDQLLLHYQQEAKEILSRGSTEAKITEVKVEPMTYLGSSQNEVLNTKPDYQKILQNQSKVFDCMELVMDELQGSVKQLQAFMDESTQCFQKVSVQLGKRSMQQLDPSPARKLLKLQLQNPPA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title