Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551742 Supplier Invitrogen™ Supplier No. PA551742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEYITKFSLE RQTWDQLLLH YQQEAKEILS RGSTEAKITE VKVEPMTYLG SSQNEVLNTK PDYQKILQNQ SKVFDCMELV MDELQGSVKQ LQAFMDESTQ CFQKVSVQLG KRSMQQLDPS PARKLLKLQL QNPPA Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

Part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DSN1
Gene Accession No. Q9H410
Gene Alias 1700022L09Rik; AW552447; C20orf172; dJ469A13.2; Dsn1; DSN1 homolog, MIS12 kinetochore complex component; DSN1, MIND kinetochore complex component, homolog; DSN1, MIND kinetochore complex component, homolog (S. cerevisiae); DSN1, MIS12 kinetochore complex component; hKNL-3; kinetochore null 3 homolog; kinetochore-associated protein DSN1 homolog; KNL3; MIS13; RP3-469A13.1
Gene Symbols DSN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DSN1. Recombinant protein control fragment (Product #RP-102190).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79980
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.