Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSPG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558297
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558297 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558297 Supplier Invitrogen™ Supplier No. PA558297
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRDNKIRQLP ELPTTLTFID ISNNRLGRKG IKQEAFKDMY DLHHLYLTDN NLDHIPLPLP ENLRALHLQN NNILEMHEDT FCNV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

The DSPG3 gene is not well-characterized and does not appear in standard genomic databases as a distinct gene or protein coding entity. It is possible that DSPG3 refers to a specific type of dermatan sulfate proteoglycan, given the acronym DSPG, which typically relates to proteoglycans involved in extracellular matrix structure and modulation. Proteoglycans like dermatan sulfate proteoglycans are pivotal in maintaining tissue resilience and regulating cell signaling pathways by interacting with growth factors and cytokines. They contribute to developmental processes and tissue repair mechanisms, playing roles in cellular adhesion, proliferation, and migration. In the context of skeletal and connective tissues, DSPGs influence matrix assembly and modulate biomechanical properties, impacting processes such as angiogenesis and fibrosis. Proteoglycans are known for their importance in disease states, including cancer and fibrosis, where altered expression or structure can affect cellular interactions and disease progression. Further research is needed to understand the specifics of DSPG3's functions and its implications in health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSPG3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPYC
Gene Accession No. Q99645
Gene Alias Dermatan sulfate proteoglycan 3; dermatan sulphate proteoglycan 3; Dspg3; DSPG3proteoglycan-lb; Epiphycan; epiphycan proteoglycan; EPYC; Pglb; PG-Lb; Proteoglycan-Lb; SLRR3B; Small chondroitin/dermatan sulfate proteoglycan
Gene Symbols EPYC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DSPG3. Recombinant protein control fragment (Product #RP-97643).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1833
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.