Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSPP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576382
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576382 Supplier Invitrogen™ Supplier No. PA576382
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes two principal proteins of the dentin extracellular matrix of the tooth. The preproprotein is secreted by odontoblasts and cleaved into dentin sialoprotein and dentin phosphoprotein. Dentin phosphoprotein is thought to be involved in the biomineralization process of dentin. Mutations in this gene have been associated with dentinogenesis imperfecta-1; in some individuals, dentinogenesis imperfecta occurs in combination with an autosomal dominant form of deafness. Allelic differences due to repeat polymorphisms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSPP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene DSPP
Gene Accession No. P97399, Q62598, Q9NZW4
Gene Alias dentin matrix protein 3; dentin phosphophoryn; Dentin phosphoprotein; dentin phosphoryn; dentin sailophosphoprotein; dentin sialophosphoprotein; Dentin sialoprotein; Dentine sialoprotein; DFNA39; DGI1; Dmp2; Dmp3; DMP-3; Dpp; DSP; DSPP; DTDP2; RDSP2
Gene Symbols DSPP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Dentin sialoprotein: 101-220.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1834, 25254, 666279
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.