Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSPP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0806 Supplier Bioss Supplier No. BS10316R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes two principal proteins of the dentin extracellular matrix of the tooth. The preproprotein is secreted by odontoblasts and cleaved into dentin sialoprotein and dentin phosphoprotein. Dentin phosphoprotein is thought to be involved in the biomineralization process of dentin. Mutations in this gene have been associated with dentinogenesis imperfecta-1; in some individuals, dentinogenesis imperfecta occurs in combination with an autosomal dominant form of deafness. Allelic differences due to repeat polymorphisms have been found for this gene.

Specifications

Antigen DSPP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DSPP
Gene Accession No. Q9NZW4
Gene Alias dentin matrix protein 3; Dentin phosphophoryn; Dentin phosphoprotein; dentin phosphoryn; dentin sailophosphoprotein; Dentin sialophosphoprotein; Dentin sialoprotein; Dentine sialoprotein; DFNA39; DGI1; Dmp2; Dmp3; DMP-3; DPP; DSP; DSPP; DTDP2; RDSP2
Gene Symbols DSPP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DSPP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1834
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.