Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DsRed-Express2 Mouse anti-Tag, Magnetic beads, Clone: OTI1C4, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA183029

Description
DsRed-Express2 fluorescent protein.Specifications
DsRed-Express2 | |
Monoclonal | |
PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide | |
Mouse | |
Affinity Chromatography | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Immunoprecipitation | |
OTI1C4 | |
DsRed-Express2 | |
Recombit protein of DsRed-Express2 expressed in HEK293 cells. | |
1 mL | |
Primary | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction