Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DsRed-Monomer Mouse anti-Tag, Clone: OTI4C7, liquid, TrueMAB™
SDP

Catalog No. 501671136
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1136 Supplier OriGene Supplier No. TA180083
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DsRed-Monomer fluorescent protein.

Specifications

Antigen DsRed-Monomer
Applications Western Blot
Classification Monoclonal
Clone OTI4C7
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Alias DsRed-Monomer
Host Species Mouse
Immunogen Recombit protein of DsRed-Monomer expressed in HEK293 cells.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.