Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dsRNA Antibody (1D3), Alexa Fluor™ 700 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320110AF700
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
dsRNA | |
Monoclonal | |
Alexa Fluor 700 | |
Double-stranded RNA | |
RDV-RNA-methylated bovine serum albumin complex. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
1D3 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
All species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction