Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dsRNA Mouse anti-Virus, Clone: 3G1, MilliporeSigma™

Mouse Monoclonal Antibody

Supplier:  MilliporeSigma MABF199625UL

 View more versions of this product

Catalog No. MABF199625


Only null left
Add to Cart

Description

Description

dsRNA Monoclonal antibody specifically detects dsRNA in Virus samples. It is validated for ELISA, Immunomicroscopy, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

dsRNA
Monoclonal
0.5 mg/mL
Purified mouse monoclonal antibody IgM in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Concentrated supernatant from C6/36 cell cultures infected with Palm Creek virus
25 μL
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
IgM κ
ELISA, Immunomicroscopy, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
3G1
Unconjugated
Mouse
Ligatrap™
RUO
Virus
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.