Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ dsRNA Chimeric Recombinant Mouse Monoclonal Antibody (1D3)
GREENER_CHOICE

Catalog No. PIMA547972
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547972 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547972 Supplier Invitrogen™ Supplier No. MA547972
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

dsRNA Chimeric Recombinant Monoclonal Antibody for ELISA, IHC, Western Blot

Double-stranded RNA (dsRNA) is a molecule composed of two complementary strands of RNA nucleotides arranged in a double helix structure. It plays a diverse role in various biological processes such as gene regulation, RNA interference, and viral defense mechanisms. It can also be used as a tool for studying gene function and as a potential therapeutic agent for treating diseases. However, it is important to note that dsRNA can trigger an immune response in some organisms, which can limit its use in certain applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dsRNA Chimeric
Applications ELISA, Immunohistochemistry, Western Blot
Classification Recombinant Monoclonal
Clone 1D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene EGFR
Gene Accession No. P00533
Host Species Mouse
Immunogen RDV-RNA-methylated bovine serum albumin complex.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1956
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.