Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106312
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106312 Supplier Invitrogen™ Supplier No. PA5106312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DSS1.

The product of this gene has been localized within the split hand/split foot malformation locus SHFM1 at chromosome 7. It has been proposed to be a candidate gene for the autosomal dominant form of the heterogeneous limb developmental disorder split hand/split foot malformation type 1. In addition, it has been shown to directly interact with BRCA2. It also may play a role in the completion of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DSS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SEM1
Gene Accession No. P60897, Q6ZVN7
Gene Alias 26S proteasome complex subunit DSS1; 26S proteasome complex subunit SEM1; C7orf76; Deleted in split hand/split foot protein 1; deleted in split hand/split foot protein 1 homolog; deleted in split-hand/split-foot 1; Dss1; ECD; Putative protein SEM1, isoform 2; Sem1; SHFD1; Shfdg1; Shfg; Shfm1; SHSF1; split hand/foot deleted gene 1; split hand/foot deleted protein 1; Split hand/foot deleted protein 1 homolog; split hand/foot malformation (ectrodactyly) type 1; split hand/foot malformation type 1 protein; Split hand/foot malformation type 1 protein homolog
Gene Symbols SEM1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SEM1(Accession P60896), corresponding to amino acid residues D35-R57.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20422, 680532, 7979
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.