Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DST Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576027 Supplier Invitrogen™ Supplier No. PA576027
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

As basal cells of stratified squamous epithelia begin to migrate in response to wound healing, they lose their cell-substrate adhesion junctions, the hemidesmosomes. The hemidesmosome is an adhesion structure of the epidermal-dermal junction in keratinocytes. When keratinocytes migrate laterally or upward to differentiate they must control the formation and disintegration of the hemidesmosomes. The bullous pemphigoid antigen BPAG1 is a hemidesmosomal protein of the cutaneous basement membrane zone. The primary sequence deduced from full-length human cDNAs predicts that this molecule consists of a central rod region and flanking globular domains. A neuronal isoform, BPAG1n3 is the result of differential splicing of BPAG1. BPAG1n3 is distinguished by its initial 32 amino acid residues and by the absence of the amino-terminal half of the actin-binding domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DST
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene DST
Gene Accession No. Q03001, Q91ZU6
Gene Alias 230 kDa bullous pemphigoid antigen; 230/240 kDa bullous pemphigoid antigen; 2310001O04Rik; A830042E19Rik; ah; athetoid; AW554249; BP230; BP240; BPA; Bpag; Bpag1; BPAG1-n; bullous pemphigoid antigen 1; bullous pemphigoid antigen 1, isoform 5; bullous pemphigoid antigen 1, isoforms 1/2/3/4; bullous pemphigoid antigen 1, isoforms 6/7; CATX15; CATX-15; D6S1101; DMH; DST; DT; dystonia musculorum protein; Dystonin; EBSB2; Hemidesmosomal plaque protein; HSAN6; KIAA0728; MACF2; microtubule actin cross-linking factor 2; mKIAA0728; nmf203; nmf339; trabeculin-beta
Gene Symbols DST
Host Species Rabbit
Immunogen Recombinant full length Human DST.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13518, 316313, 667
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.