Learn More
DSTN Mouse anti-Canine, Human, Clone: OTI1G8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502625

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
DSTN | |
Monoclonal | |
Unconjugated | |
DSTN | |
ACTDP, ADF, HEL32, bA462D18.2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
11034, 477147 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1G8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P60981 | |
DSTN | |
Full length human recombit protein of human DSTN produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.