Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSU Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983271
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3271 Supplier Bioss Supplier No. BS14433R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plays a role in the incorporation of pigments into hair. May function in membrane fusion and regulate the biogenesis of disk membranes of photoreceptor rod cells.

Specifications

Antigen DSU
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MREG
Gene Accession No. Q8N565
Gene Alias BOS_2061; Dilute suppressor protein; Dilute suppressor protein homolog; DSU; Gm974; HDCGA21P; LOC100347403; Melanoregulin; Mreg; RGD1562333; Wdt2; Whn-dependent transcript 2
Gene Symbols MREG
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DSU.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55686
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.