Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynex™ DSX™ Automated ELISA Processor

Catalog No. p-7147245
Click to view available options
Description:
2 plates system
4 plates system
4 plates system (with no incubator)
Incubator Module
This item is not returnable. View return policy
Appropriate precautions must be taken when working with biohazardous material.
This item is not returnable. View return policy
Appropriate precautions must be taken when working with biohazardous material.

Busy clinical and medium-throughput private and industrial labs require advanced automation that can handle virtually every ELISA immunoassay, all in a reliable, cost-effective package. Look no further than the DSX™

  • Completely automated open system
  • Well fill verification
  • Unique dispense synchronization
  • 96 on-board samples
  • Up to 12 different assays simultaneously
  • Automatic data reduction
  • Easy-to-Use Revelation DSX Software
  • Disposable sample and reagent tips
  • Specifications

    Type Open System
    Description 2 plates system
    For Use With (Application) ELISA Assays
    Sample Dilution (192) 24 x 8 deep-well strips Maximum dilution: 1 to 36,100
    Throughput Assay dependent
    Analytical Method Photometric
    Capacity 98 samples on-board
    Certifications/Compliance EN 61326-1:2013EN 61326-2-6:2013 IEC/EN 61010-1:2010 Ed.3 IEC/EN 61010-2-010:2014 Ed.3 IEC/EN 61010-2-101 issued 2015/08/14 Ed. 2 CSA C22.2 No. 61010-1:2012 Ed.3 CSA C22.2 No. 61010-2-101:2015 Ed.2 RoHS Compliant
    Includes One year warranty against defects in parts, materials, and workmanship
    Wavelength 405/690 nm
    Depth (English) 36 in.
    Depth (Metric) 91 cm
    Height (English) 32 in.
    Height (Metric) 81 cm
    Width (English) 42 in.
    Width (Metric) 106 cm
    Electrical Requirements 100/240 V auto-switching
    Frequency 50/60 Hz
    Data Management Features Revelation DSX™ Software
    Operating System Windows 7/ Windows 10
    Measurement Method Absorbance
    Weight (English) 537 lb.
    Weight (Metric) 244 kg
    Show More Show Less

    DYNEX makes no representations, warranties or performance claims with respect to the performance of Agility for any specific application, including clinical application or for the use of the Agility system with any reagents, assays or other products.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.