Learn More
Tocris Bioscience™ dTAG-7

Description
FKBP12F36V can be expressed as a fusion with a target protein of interest using genome engineering techniques, via transgene expression or CRISPR-mediated locus-specific knock-in. Custom knock-in cell lines for the dTAG and aTAG platforms are available from our sister brand R&D Systems . Email TPD@bio-techne.com to enquire.
Plasmid vectors for the lentiviral expression and CRISPR-mediated knock-in of FKBP12F36V are available from Addgene.
Specifications
Specifications
| Chemical Name or Material | (1 R)-3-(3,4-Dimethoxyphenyl)-1-(2-((19-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)-2,18-dioxo-7,10,13-trioxa-3,17-diazanonadecyl)oxy)phenyl)propyl (2 S)-1-(-2-(3,4,5-trimethoxyphenyl)butanoyl)piperidine-2-carboxylate |
| CAS | 2064175-32-0 |
| Quantity | 5 mg |
| Target | TAG Degradation Platforms |
| Molecular Formula | C63H79N5O19 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.