Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DTD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | DTD1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
DTD1 Polyclonal specifically detects DTD1 in Human samples. It is validated for Western Blot.Specifications
DTD1 | |
Polyclonal | |
Rabbit | |
Q8TEA8 | |
92675 | |
Synthetic peptide directed towards the C-terminal region of Human DTD1. Located within the following region: APGTATSDPKQLSKLEKQQQRKEKTRAKGPSESSKERNTPRKEDRSASSG | |
Primary | |
23 kDa |
Western Blot | |
Unconjugated | |
RUO | |
bA379J5.3, bA555E18.1, C20orf88HARS2, chromosome 20 open reading frame 88, DNA-unwinding element-binding protein, D-tyrosyl-tRNA deacylase 1 homolog (S. cerevisiae), D-tyrosyl-tRNA(Tyr) deacylase 1, DUEB, EC 3.1, Histidyl-tRNA synthase-related, histidyl-tRNA synthetase 2, MGC119131, MGC41905, pqn-68 | |
DTD1 | |
IgG | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title