Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DTD1 Antibody (OTI1E1), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272335C
Description
DTD1 Monoclonal specifically detects DTD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
DTD1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DTD1 | |
Full length recombinant protein of human DTD1 (NP_543010) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1E1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
bA379J5.3, bA555E18.1, C20orf88HARS2, chromosome 20 open reading frame 88, DNA-unwinding element-binding protein, D-tyrosyl-tRNA deacylase 1 homolog (S. cerevisiae), D-tyrosyl-tRNA(Tyr) deacylase 1, DUEB, EC 3.1, Histidyl-tRNA synthase-related, histidyl-tRNA synthetase 2, MGC119131, MGC41905, pqn-68 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
92675.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction