Learn More
DTD1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304671A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Bovine
An aminoacyl-tRNA editing enzyme that deacylates mischarged D-aminoacyl-tRNAs. Also deacylates mischarged glycyl-tRNA(Ala), protecting cells against glycine mischarging by AlaRS. Acts via tRNA-based rather than protein-based catalysis; rejects L-amino acids rather than detecting D-amino acids in the active site. By recycling D-aminoacyl-tRNA to D-amino acids and free tRNA molecules, this enzyme counteracts the toxicity associated with the formation of D-aminoacyl-tRNA entities in vivo and helps enforce protein L-homochirality. Possible ATPase involved in DNA replication, may facilitate loading of CDC45 onto pre-replication complexes.Specifications
DTD1 | |
Polyclonal | |
Unconjugated | |
DTD1 | |
bA379J5.3, bA555E18.1, C20orf88, chromosome 20 open reading frame 88, DNA-unwinding element-binding protein B, D-tyrosyl-tRNA deacylase 1 homolog, D-tyrosyl-tRNA deacylase 1 homolog (S. cerevisiae), DUEB, DUE-B, HARS2, histidyl-tRNA synthase-related, histidyl-tRNA synthetase 2, MGC119131, MGC41905, pqn-68 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92675 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8TEA8 | |
DTD1 | |
Between 159 and 209 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.