Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dtk/TYRO3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$181.90 - $402.00

Specifications

Antigen Dtk/TYRO3
Dilution Immunohistochemistry-Paraffin 1: 100
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP223725SS
SDP
View Documents
Novus Biologicals
NBP223725SS
0.025 mL
Each for $181.90
Only null left
Add to Cart
 
NBP223725
SDP
View Documents
Novus Biologicals
NBP223725
0.1 mL
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Dtk/TYRO3 Polyclonal specifically detects Dtk/TYRO3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Dtk/TYRO3
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases
0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
TYRO3
IgG
Unpurified
The amino acid sequence used as immunogen is 100% homologous in human and 81% homologous in chicken.
Immunohistochemistry-Paraffin 1: 100
Polyclonal
Antisera
RUO
Human
Q06418
7301
Within the range of amino acids 110-140 of human TYRO3 protein were used as the immunogen.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.