Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dtk/TYRO3 Antibody, PE/Atto594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128635PEATT594
Description
Dtk/TYRO3 Polyclonal specifically detects Dtk/TYRO3 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).Specifications
Dtk/TYRO3 | |
Polyclonal | |
Flow Cytometry, Flow (Intracellular) | |
Brt, BYK, DTK, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY | |
Rabbit | |
Affinity Purified | |
Cancer, Neuroscience, Neurotransmission, Protein Kinase, Tyrosine Kinases | |
7301.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
TYRO3 | |
Synthetic peptide made to an internal portion of the human TYRO3 protein (within residues 100-250). [Swiss-Prot No. Q06418] | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction