Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588380 Supplier Invitrogen™ Supplier No. PA588380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEACSESRNR VKRRLDSSCL ESVKQKCVKS CNCVTELDGQ VENLHLDLCC LAGNQEDLSK DSLGPTKSSK IEGAGTSISE PPSPISPYAS ESCGTLPLPL RPCGEGSEMV GKENSSPENK NWLLAMAAKR KAENPSPRSP SSQTPNSRRQ SGKTLPSPVT ITPSSMRKIC TYFHRKSQED FCGPEHSTEL; Positive Samples: Mouse testis, Mouse liver, Rat brain; Cellular Location: Chromosome, Cytoplasm, Nucleoplasmic side, Nucleus, Nucleus membrane, Peripheral membrane protein, centrosome, cytoskeleton, microtubule organizing center.

Substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control, DNA damage response and translesion DNA synthesis. The DCX(DTL) complex, also named CRL4(CDT2) complex, mediates the polyubiquitination and subsequent degradation of CDT1 and CDKN1A/p21(CIP1). CDT1 degradation in response to DNA damage is necessary to ensure proper cell cycle regulation of DNA replication. CDKN1A/p21(CIP1) degradation during S phase or following UV irradiation is essential to control replication licensing. Most substrates require their interaction with PCNA for their polyubiquitination: substrates interact with PCNA via their PIP-box, and those containing the 'K+4' motif in the PIP box, recruit the DCX(DTL) complex, leading to their degradation. In undamaged proliferating cells, the DCX(DTL) complex also promotes the 'Lys-164' monoubiquitination of PCNA, thereby being involved in PCNA-dependent translesion DNA synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DTL
Gene Accession No. Q3TLR7
Gene Alias 2810047L02Rik; 5730564G15Rik; CDT2; CDW1; DCAF2; DDB1 and CUL4 associated factor 2; DDB1- and CUL4-associated factor 2; denticleless E3 ubiquitin protein ligase homolog; denticleless E3 ubiquitin protein ligase homolog (Drosophila); denticleless homolog; denticleless homolog (Drosophila); denticleless protein homolog; DTL; L2dtl; lethal(2) denticleless protein homolog; Meth A RAMP; Meth A retinoic acid-regulated nuclear matrix-associated protein; RA regulated nuclear matrix associated protein; RAMP; RA-regulated nuclear matrix-associated protein; Retinoic acid-regulated nuclear matrix-associated protein; RGD1310439
Gene Symbols DTL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 541-730 of human DTL (NP_0575323).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305073, 76843
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.